• ホーム
  • 検索結果
  • Decitabine inhibits tumor cell proliferation and up-regulates e-cadherin expression in Epstein-Barr virus-associated gastric cancer.
Decitabine inhibits tumor cell proliferation and up-regulates e-cadherin expression in Epstein-Barr virus-associated gastric cancer.
Journal of medical virology (2016-07-20)
Munetaka Nakamura, Jun Nishikawa, Mari Saito, Kouhei Sakai, Sho Sasaki, Shinichi Hashimoto, Takeshi Okamoto, Yutaka Suehiro, Takahiro Yamasaki, Isao Sakaida
要旨
The present study investigated the effect of a DNA demethylating agent, decitabine, against Epstein-Barr virus-associated gastric cancer (EBVaGC). Decitabine inhibited cell growth and induced G2/M arrest and apoptosis in EBVaGC cell lines. The expression of E-cadherin was up-regulated and cell motility was significantly inhibited in the cells treated with decitabine. The promoter regions of p73 and RUNX3 were demethylated, and their expression was up-regulated by decitabine. They enhanced the transcription of p21, which induced G2/M arrest and apoptosis through down-regulation of c-Myc. Decitabine also induced the expression of BZLF1 in SNU719. Induction of EBV lytic infection was an alternative way to cause apoptosis of the host cells. This study is the first report to reveal the effectiveness of a demethylating agent in inhibiting tumor cell proliferation and up-regulation of E-cadherin in EBVaGC. J. Med. Virol. 89:508-517, 2017. © 2016 Wiley Periodicals, Inc.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
α,α-Dicyanoethyl acetate, 98%