• ホーム
  • 検索結果
  • Teriflunomide restores 5-azacytidine sensitivity via activation of pyrimidine salvage in 5-azacytidine-resistant leukemia cells.
Teriflunomide restores 5-azacytidine sensitivity via activation of pyrimidine salvage in 5-azacytidine-resistant leukemia cells.
Oncotarget (2017-10-21)
Satoshi Imanishi, Ryoko Takahashi, Seiichiro Katagiri, Chiaki Kobayashi, Tomohiro Umezu, Kazuma Ohyashiki, Junko H Ohyashiki
要旨
Previous studies showed that downregulation of pyrimidine salvage underlies resistance against 5-azacytidine (AZA), indicating an important role for
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
α,α-Dicyanoethyl acetate, 98%