  • Search Results
  • A role for TSPO in mitochondrial Ca2+ homeostasis and redox stress signaling.
A role for TSPO in mitochondrial Ca2+ homeostasis and redox stress signaling.
Cell death & disease (2017-06-24)
Jemma Gatliff, Daniel A East, Aarti Singh, Maria Soledad Alvarez, Michele Frison, Ivana Matic, Caterina Ferraina, Natalie Sampson, Federico Turkheimer, Michelangelo Campanella
ABSTRACT
The 18 kDa translocator protein TSPO localizes on the outer mitochondrial membrane (OMM). Systematically overexpressed at sites of neuroinflammation it is adopted as a biomarker of brain conditions. TSPO inhibits the autophagic removal of mitochondria by limiting PARK2-mediated mitochondrial ubiquitination via a peri-organelle accumulation of reactive oxygen species (ROS). Here we describe that TSPO deregulates mitochondrial Ca2+ signaling leading to a parallel increase in the cytosolic Ca2+ pools that activate the Ca2+-dependent NADPH oxidase (NOX) thereby increasing ROS. The inhibition of mitochondrial Ca2+ uptake by TSPO is a consequence of the phosphorylation of the voltage-dependent anion channel (VDAC1) by the protein kinase A (PKA), which is recruited to the mitochondria, in complex with the Acyl-CoA binding domain containing 3 (ACBD3). Notably, the neurotransmitter glutamate, which contributes neuronal toxicity in age-dependent conditions, triggers this TSPO-dependent mechanism of cell signaling leading to cellular demise. TSPO is therefore proposed as a novel OMM-based pathway to control intracellular Ca2+ dynamics and redox transients in neuronal cytotoxicity.
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
イオノマイシン カルシウム塩, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO
Sigma-Aldrich
エチレンジアミン四酢酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
テトラメチルローダミンメチルエステルパークロレート, ≥95%