  • Search Results
  • Syntaphilin controls a mitochondrial rheostat for proliferation-motility decisions in cancer.
Syntaphilin controls a mitochondrial rheostat for proliferation-motility decisions in cancer.
The Journal of clinical investigation (2017-09-12)
M Cecilia Caino, Jae Ho Seo, Yuan Wang, Dayana B Rivadeneira, Dmitry I Gabrilovich, Eui Tae Kim, Ashani T Weeraratna, Lucia R Languino, Dario C Altieri
ABSTRACT
Tumors adapt to an unfavorable microenvironment by controlling the balance between cell proliferation and cell motility, but the regulators of this process are largely unknown. Here, we show that an alternatively spliced isoform of syntaphilin (SNPH), a cytoskeletal regulator of mitochondrial movements in neurons, is directed to mitochondria of tumor cells. Mitochondrial SNPH buffers oxidative stress and maintains complex II-dependent bioenergetics, sustaining local tumor growth while restricting mitochondrial redistribution to the cortical cytoskeleton and tumor cell motility. Conversely, introduction of stress stimuli to the microenvironment, including hypoxia, acutely lowered SNPH levels, resulting in bioenergetics defects and increased superoxide production. In turn, this suppressed tumor cell proliferation but increased tumor cell invasion via greater mitochondrial trafficking to the cortical cytoskeleton. Loss of SNPH or expression of an SNPH mutant lacking the mitochondrial localization sequence resulted in increased metastatic dissemination in xenograft or syngeneic tumor models in vivo. Accordingly, tumor cells that acquired the ability to metastasize in vivo constitutively downregulated SNPH and exhibited higher oxidative stress, reduced cell proliferation, and increased cell motility. Therefore, SNPH is a stress-regulated mitochondrial switch of the cell proliferation-motility balance in cancer, and its pathway may represent a therapeutic target.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
モノクローナル抗FLAG® M2抗体 マウス宿主抗体, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
モノクロナール抗β-アクチン マウス宿主抗体, clone AC-15, ascites fluid
Sigma-Aldrich
カルボニルシアニド 4-(トリフルオロメトキシ)フェニルヒドラゾン, ≥98% (TLC), powder
Sigma-Aldrich
ヘマトキシリン溶液、マイヤー法
Sigma-Aldrich
モノクロナール抗β-チューブリン マウス宿主抗体, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
2,3-ジメトキシ-1,4-ナフトキノン, ≥99%, solid
Sigma-Aldrich
Anti-Paxillin Antibody, clone 5H11, clone 5H11, Upstate®, from mouse
Sigma-Aldrich
アミロイドタンパク質非Aβ成分, ≥80% (HPLC)