Merck
Regulation of GLUT4 activity in myotubes by 3-O-methyl-d-glucose.
Biochimica et biophysica acta (2017-06-27)
Ofer Shamni, Guy Cohen, Arie Gruzman, Hilal Zaid, Amira Klip, Erol Cerasi, Shlomo Sasson
ABSTRACT
The rate of glucose influx to skeletal muscles is determined primarily by the number of functional units of glucose transporter-4 (GLUT4) in the myotube plasma membrane. The abundance of GLUT4 in the plasma membrane is tightly regulated by insulin or contractile activity, which employ distinct pathways to translocate GLUT4-rich vesicles from intracellular compartments. Various studies have indicated that GLUT4 intrinsic activity is also regulated by conformational changes and/or interactions with membrane components and intracellular proteins in the vicinity of the plasma membrane. Here we show that the non-metabolizable glucose analog 3-O-methyl-d-glucose (MeGlc) augmented the rate of hexose transport into myotubes by increasing GLUT4 intrinsic activity without altering the content of the transporter in the plasma membrane. This effect was not a consequence of ATP depletion or hyperosmolar stress and did not involve Akt/PKB or AMPK signal transduction pathways. MeGlc reduced the inhibitory potency (increased K
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder