  • Search Results
  • A Novel Phenylchromane Derivative Increases the Rate of Glucose Uptake in L6 Myotubes and Augments Insulin Secretion from Pancreatic Beta-Cells by Activating AMPK.
A Novel Phenylchromane Derivative Increases the Rate of Glucose Uptake in L6 Myotubes and Augments Insulin Secretion from Pancreatic Beta-Cells by Activating AMPK.
Pharmaceutical research (2017-10-07)
Naomi Rozentul, Yosef Avrahami, Moran Shubely, Laura Levy, Anna Munder, Guy Cohen, Erol Cerasi, Shlomo Sasson, Arie Gruzman
ABSTRACT
A series of novel polycyclic aromatic compounds that augment the rate of glucose uptake in L6 myotubes and increase glucose-stimulated insulin secretion from beta-cells were synthesized. Designing these molecules, we have aimed at the two main pathogenic mechanisms of T2D, deficient insulin secretion and diminished glucose clearance. The ultimate purpose of this work was to create a novel antidiabetic drug candidate with bi-functional mode of action. All presented compounds were synthesized, and characterized in house. INS-1E cells and L6 myoblasts were used for the experiments. The rate of glucose uptake, mechanism of action, level of insulin secretion and the druggability of the lead compound were studied. The lead compound (6-(1,3-dithiepan-2-yl)-2-phenylchromane), dose- and time-dependently at the low μM range increased the rate of glucose uptake in L6 myotubes and insulin secretion in INS-1E cells. The compound exerted its effects through the activation of the LKB1 (Liver Kinase B1)-AMPK pathway. In vitro metabolic parameters of this lead compound exhibited good druggability. We anticipate that bi-functionality (increased rate of glucose uptake and augmented insulin secretion) will allow the lead compound to be a starting point for the development of a novel class of antidiabetic drugs.
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder