• ホーム
  • 検索結果
  • PAX9 regulates squamous cell differentiation and carcinogenesis in the oro-oesophageal epithelium.
PAX9 regulates squamous cell differentiation and carcinogenesis in the oro-oesophageal epithelium.
The Journal of pathology (2017-10-22)
Zhaohui Xiong, Shuang Ren, Hao Chen, Yao Liu, Caizhi Huang, Yawan Lyvia Zhang, Joab Otieno Odera, Tong Chen, Ralf Kist, Heiko Peters, Katherine Garman, Zheng Sun, Xiaoxin Chen
要旨
PAX9 is a transcription factor of the PAX family characterized by a DNA-binding paired domain. Previous studies have suggested a potential role of PAX9 in squamous cell differentiation and carcinogenesis of the oro-oesophageal epithelium. However, its functional roles in differentiation and carcinogenesis remain unclear. In this study, Pax9 deficiency in mouse oesophagus promoted cell proliferation, delayed cell differentiation, and altered the global gene expression profile. Ethanol exposure downregulated PAX9 expression in human oesophageal epithelial cells in vitro and mouse forestomach and tongue in vivo. We further showed that PAX9 was downregulated in human oro-oesophageal squamous cell carcinoma (OESCC), and its downregulation was associated with alcohol drinking and promoter hypermethylation. Moreover, ad libitum feeding with a liquid diet containing ethanol for 40 weeks or Pax9 deficiency promoted N-nitrosomethylbenzylamine-induced squamous cell carcinogenesis in mouse tongue, oesophagus, and forestomach. In conclusion, PAX9 regulates squamous cell differentiation in the oro-oesophageal epithelium. Alcohol drinking and promoter hypermethylation are associated with PAX9 silencing in human OESCC. PAX9 downregulation may contribute to alcohol-associated oro-oesophageal squamous cell carcinogenesis. Copyright © 2017 Pathological Society of Great Britain and Ireland. Published by John Wiley & Sons, Ltd.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
モノクロナール抗増殖細胞核抗原 マウス宿主抗体, clone PC 10, ascites fluid
Sigma-Aldrich
α,α-Dicyanoethyl acetate, 98%