製品
製品アプリケーションサービスリソースサポート
[ログイン / 登録](https://www.sigmaaldrich.com/oidc-sign-in)
[注文検索](https://www.sigmaaldrich.com/JP/ja/order-lookup)
[クイックオーダー](https://www.sigmaaldrich.com/JP/ja/quick-order)
カート0
[ホーム](https://www.sigmaaldrich.com/JP/ja)[組織工学](https://www.sigmaaldrich.com/JP/ja/applications/materials-science-and-engineering/tissue-engineering)メソポーラス材料
# メソポーラス材料
メソポーラス分子ふるいとしても知られるメソポーラス材料は、メソスケール(直径2~50 nm)の細孔を持つ3次元ナノ構造体の一種であり、1000 m2/gを超える表面積を示す多孔質材料です1 。特徴的な(場合によっては)秩序性を有する構造という点では、結晶性ゼオライトと、他のタイプの3次元構造体材料(たとえば100 nmを超える構造を持つ3DOM(3D ordered mesoporous)や直接描画法による材料など)との間のユニークな位置を占めています。メソポーラス材料は、自己組織化プロセスによって、ゾル-ゲル前駆体(金属アルコキシドなど)と構造規定性両親媒性物質(通常はブロックコポリマーや界面活性剤)の混合液から形成されます(__図1__)2,3。自己組織化鋳型法による「ワンポット合成」は柔軟性が高いため細孔のサイズと3次元形状(中間相)を同時に制御することができます。さらに、有機シロキサン(RSi(OR’)3)やビス(有機シロキサン)((R’O)3Si-R-Si(OR’)3)など、有機官能基で修飾された前駆体を最初の反応混合物に加えることで、細孔表面の官能性を変更することもできます4 。他方、自己組織化構造の規則性や配向性を広い範囲で制御することは比較的難しいため、この種の材料は通常、3DOM材料や直接描画材料と比較すると、欠陥が多く、構造正確性が劣ります。
__図1__メソポーラスシリカ(MCM-41)の合成経路の模式図2。(I)臭化セチルトリメチルアンモニウム(CTAB、製品番号:[52370](https://www.sigmaaldrich.com/product/aldrich/52370))などの界面活性剤を用いて水中に液晶ミセルを形成。(II)テトラエチルオルトケイ酸(TEOS、製品番号:[131903](https://www.sigmaaldrich.com/product/aldrich/131903))などのセラミックゾル-ゲル前駆体をこのミセル溶液に加えると、加水分解および縮合を起こし、ミセル周辺にシリカのネットワークが形成される。(III)熱処理(焼成)または溶媒抽出で有機鋳型を除去すると、メソポーラスなセラミック材料(この場合は六方晶系秩序構造MCM-41シリカ)の骨格が得られる。
このように、3次元構造の作製方法によって得られる材料の利点が異なるため、最適な用途がそれぞれ決まってきます。「柔らかい」両親媒性物質の鋳型で作られた秩序的なメソポーラス材料は、ゼオライトのもつ細孔サイズの制約を克服し、かさ高い分子を拡散しやすくします。この材料は、広い範囲における材料の秩序性がそれほど重視されない触媒技術や吸着技術に向いています。たとえば、酸性アルミノケイ酸塩は、流動接触分解(FCC:Fluid Catalytic Craking)や凝縮媒体中での化学変換プロセスへの利用について研究が進められています5 。表面に官能基を導入したメソポーラス分子ふるいはセンサーの活性素子として用いられます6 。色素などの大型の光学活性分子7 (例:ローダミン6G、製品番号: [252433](https://www.sigmaaldrich.com/JP/ja/product/aldrich/252433))や共役ポリマー8 (例:MEH-PPV、 製品番号: [541443](https://www.sigmaaldrich.com/JP/ja/product/aldrich/541443))をメソ細孔に組み込んで、独特の光電子特性を有する複合材料を作ることもできます。
アルカリ水熱条件下におけるメソポーラスケイ酸塩の作製には、カチオン性四級アンモニウム系界面活性剤がしばしば用いられます。一方、メソポーラスアルミナの水系合成や、正電荷の対イオンまたは構造規定性の補助剤を追加したアルカリ性条件下での合成には、アニオン性界面活性剤が用いられます9,10 。また、秩序性のないワームホール型(虫食い)シリカ(HMS、MSU)の調製や、酸性条件下での規則性構造を持つシリカの合成には、非イオン性界面活性剤を用いることもできます3 。細孔径が5 nm以上で均一な秩序性の高いメソポーラス材料は、酸性の水性媒体中で、PEG-PPG-PEG(プルロニック)トリブロックコポリマーを鋳型として用いて作製できます11。
## 関連製品
申し訳ございませんが、想定外のエラーが発生しました。
Response not successful: Received status code 500
### 参考文献
1\.
Beck JS, Vartuli JC. 1996. Recent advances in the synthesis, characterization and applications of mesoporous molecular sieves. Current Opinion in Solid State and Materials Science. 1(1):76-87. [https://doi.org/10.1016/s1359-0286(96)80014-3](https://doi.org/10.1016/s1359-0286%2896%2980014-3)
2\.
Beck JS, Vartuli JC, Roth WJ, Leonowicz ME, Kresge CT, Schmitt KD, Chu CTW, Olson DH, Sheppard EW, McCullen SB, et al. 1992. A new family of mesoporous molecular sieves prepared with liquid crystal templates. J. Am. Chem. Soc.. 114(27):10834-10843. [https://doi.org/10.1021/ja00053a020](https://doi.org/10.1021/ja00053a020)
3\.
Wan Y, Zhao. 2007. On the Controllable Soft-Templating Approach to Mesoporous Silicates. Chem. Rev.. 107(7):2821-2860. [https://doi.org/10.1021/cr068020s](https://doi.org/10.1021/cr068020s)
4\.
Young SK. 2006. Sol-Gel Science for Ceramic Materials Material Matters .(1):8-13.
5\.
Liu Y, Pinnavaia TJ. 2002. Aluminosilicate mesostructures with improved acidity and hydrothermal stability. J. Mater. Chem.. 12(11):3179-3190. [https://doi.org/10.1039/b204094h](https://doi.org/10.1039/b204094h)
6\.
Wirnsberger G, Scott BJ, Stucky GD. 2001. pH Sensing with mesoporous thin films. Chem. Commun..(1):119-120. [https://doi.org/10.1039/b003995k](https://doi.org/10.1039/b003995k)
7\.
Yang P. 2000. Mirrorless Lasing from Mesostructured Waveguides Patterned by Soft Lithography. 287(5452):465-467. [https://doi.org/10.1126/science.287.5452.465](https://doi.org/10.1126/science.287.5452.465)
8\.
Nguyen T, Wu J, Tolbert S, Schwartz B. 2001. Control of Energy Transport in Conjugated Polymers Using an Ordered Mesoporous Silica Matrix. Adv. Mater.. 13(8):609-611. [https://doi.org/10.1002/1521-4095(200104)13:8<609::AID-ADMA609>3.0.CO;2-%23](https://doi.org/10.1002/1521-4095%28200104%2913:8%3C609::AID-ADMA609%3E3.0.CO;2-%23)
9\.
Huo Q, Margolese DI, Ciesla U, Feng P, Gier TE, Sieger P, Leon R, Petroff PM, Schüth F, Stucky GD. 1994. Generalized synthesis of periodic surfactant/inorganic composite materials. Nature. 368(6469):317-321. [https://doi.org/10.1038/368317a0](https://doi.org/10.1038/368317a0)
10\.
Che S, Garcia-Bennett AE, Yokoi T, Sakamoto K, Kunieda H, Terasaki O, Tatsumi T. 2003. A novel anionic surfactant templating route for synthesizing mesoporous silica with unique structure. Nature Mater. 2(12):801-805. [https://doi.org/10.1038/nmat1022](https://doi.org/10.1038/nmat1022)
11\.
Zhao D, Huo Q, Feng J, Chmelka BF, Stucky GD. 1998. Nonionic Triblock and Star Diblock Copolymer and Oligomeric Surfactant Syntheses of Highly Ordered, Hydrothermally Stable, Mesoporous Silica Structures. J. Am. Chem. Soc.. 120(24):6024-6036. [https://doi.org/10.1021/ja974025i](https://doi.org/10.1021/ja974025i)
__関連資料__
- [メソポーラス材料:特徴・評価・応用](https://www.sigmaaldrich.com/JP/ja/technical-documents/technical-article/materials-science-and-engineering/nanoparticle-and-microparticle-synthesis/mesoporous-materials)
- [3Dバイオプリンティング:組織および疾患モデルへの応用](https://www.sigmaaldrich.com/JP/ja/technical-documents/technical-article/materials-science-and-engineering/3d-bioprinting/3d-bioprinting-tissue)
- [バイオプリンティング―生体組織工学および再生医療に向けて―](https://www.sigmaaldrich.com/JP/ja/technical-documents/technical-article/materials-science-and-engineering/3d-bioprinting/bioprinting-for-tissue-engineering)
- [光架橋性ゼラチンハイドロゲル:高分解能3Dプリンティング向けの多用途性材料](https://www.sigmaaldrich.com/JP/ja/technical-documents/technical-article/materials-science-and-engineering/3d-bioprinting/photo-crosslinkable-gelatin-hydrogel)
- [3Dプリンティンググラフェンインク:電子的および生物医学的構造およびデバイスの作製](https://www.sigmaaldrich.com/JP/ja/technical-documents/technical-article/materials-science-and-engineering/3d-printing/3d-printing-graphene-ink)
- [キトサンを基盤とした生体材料](https://www.sigmaaldrich.com/JP/ja/technical-documents/technical-article/materials-science-and-engineering/drug-delivery/chitosan-based-biomaterials)
- [生体材料科学における「クリック」ケミストリー](https://www.sigmaaldrich.com/JP/ja/technical-documents/technical-article/materials-science-and-engineering/drug-delivery/click-chemistry)
- [デンドロンおよび超分岐ポリマー](https://www.sigmaaldrich.com/JP/ja/technical-documents/technical-article/materials-science-and-engineering/drug-delivery/dendrimers-dendrons)
- [関連資料をもっと見る](https://www.sigmaaldrich.com/JP/ja/search/facet-search?focus=sitecontent&term=facet-search)
__関連製品分野__
- [酸化物・セラミックス](https://www.sigmaaldrich.com/JP/ja/products/materials-science/energy-materials/oxides-and-ceramics)
- [バイオメディカルポリマー](https://www.sigmaaldrich.com/JP/ja/products/materials-science/biomedical-materials/biomedical-polymers)
- [無機・金属ナノ材料](https://www.sigmaaldrich.com/JP/ja/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
- [有機EL(OLED・PLED)材料](https://www.sigmaaldrich.com/JP/ja/products/materials-science/electronic-materials/oled-and-pled-materials)
- [フォトニック・光学材料](https://www.sigmaaldrich.com/JP/ja/products/materials-science/electronic-materials/photonic-and-optical-materials)
- [サステナビリティの要素を備えた製品](https://www.sigmaaldrich.com/JP/ja/products/chemistry-and-biochemicals/greener-alternative-products)
- [電池材料](https://www.sigmaaldrich.com/JP/ja/products/materials-science/energy-materials/battery-materials)
TOP
__ログインして続行__
続きを確認するには、ログインするか、新規登録が必要です。
ログイン__アカウントをお持ちではありませんか?__お客様情報を登録
サポート[お問い合わせ](https://www.sigmaaldrich.com/JP/ja/support/customer-support)[各国の連絡先](https://www.sigmaaldrich.com/JP/ja/collections/offices)[Eコマース FAQ](https://www.sigmaaldrich.com/JP/ja/life-science/ecommerce/ecommerce-faq)[安全データシート (SDS)](https://www.sigmaaldrich.com/JP/ja/documents-search?tab=sds)[製品のCOA/COO](https://www.sigmaaldrich.com/JP/ja/documents-search?tab=coa)[品質・規制](https://www.sigmaaldrich.com/JP/ja/life-science/quality-and-regulatory-management)[計算ツール・アプリ](https://www.sigmaaldrich.com/JP/ja/support/calculators-and-apps)[ウェビナー[英語]](https://www.sigmaaldrich.com/JP/ja/collections/webinars)
オーダー[クイックオーダー](https://www.sigmaaldrich.com/JP/ja/quick-order)[カスタム製品](https://www.sigmaaldrich.com/JP/ja/services/custom-products)[Eコマースソリューション](https://www.sigmaaldrich.com/JP/ja/life-science/ecommerce/ecommerce-solutions)
会社情報[メルクについて](https://www.sigmaaldrich.com/JP/ja/life-science/about-us)[企業責任](https://www.sigmaaldrich.com/JP/ja/life-science/ssbi)[イベント](https://www.sigmaaldrich.com/JP/ja/collections/events)[プレスリリース](https://www.sigmaaldrich.com/JP/ja/collections/press)[プログラム](https://www.sigmaaldrich.com/JP/ja/life-science/partnership-programs)[採用情報](https://careers.merckgroup.com/global/en)[拠点](https://www.sigmaaldrich.com/JP/ja/collections/offices)
ソーシャルメディア
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
Merck
__研究、開発、製造__
私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。
[](https://www.sigmaaldrich.com/JP/ja)
[Sigma-Aldrich®ソリューション](https://www.sigmaaldrich.com/JP/ja/life-science/sigma-aldrich)[BioReliance®ソリューション](https://www.sigmaaldrich.com/JP/ja/life-science/bioreliance)[Millipore®ソリューション](https://www.sigmaaldrich.com/JP/ja/life-science/millipore)[SAFC®ソリューション](https://www.sigmaaldrich.com/JP/ja/life-science/safc)[Milli-Q®ソリューション](https://www.sigmaaldrich.com/JP/ja/life-science/milliq)[Supelco®ソリューション](https://www.sigmaaldrich.com/JP/ja/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。
[サイト利用規約](https://www.sigmaaldrich.com/JP/ja/life-science/legal/site-use-terms)|[個人情報保護方針](https://www.sigmaaldrich.com/JP/ja/life-science/legal/privacy-statement)|[一般販売条件](https://www.sigmaaldrich.com/JP/ja/life-science/legal/terms-and-conditions)|[著作権に関する許諾](https://www.sigmaaldrich.com/JP/ja/life-science/legal/copyright-consent)|[サイトマップ](https://www.sigmaaldrich.com/JP/ja/site-map)
An unknown error has occured.