ALNH9AA07F10
Aladdin Scientific
Sarcosine anhydride
mp
145-147 C
Iupac Name
1,4-dimethyl-2,5-piperazinedione
Inchi Code
1S/C6H10N2O2/c1-7-3-6(10)8(2)4-5(7)9/h3-4H2,1-2H3
InChI key
PKDGRAULLDDTRN-UHFFFAOYSA-N
storage temp.
Room temperature
Purity
98%
Country of Origin
US
Shipping Temperature
Normal
Product Link
https://www.aladdinsci.com/sarcosine-anhydride-bioactive-small-molecules-by-aladdin-scientific-s358734.html
Melting Point
145-147 C
Precautionary statements
MSDS
https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/S358734-SCI_eaa12645576a81f443467b18f49806b9.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service