Merck
5-FORMYL-2,4,6-TRIMETHYL-NICOTINIC ACID ETHYL ESTER AldrichCPR

5-FORMYL-2,4,6-TRIMETHYL-NICOTINIC ACID ETHYL ESTER

5-FORMYL-2,4,6-TRIMETHYL-NICOTINIC ACID ETHYL ESTER AldrichCPR
Linear Formula:
C12H15NO3
CAS Number:
Molecular Weight:
221.258

Products

Formula Weight
Special Grade

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-2 of 2