Merck
Urine-based detection of intestinal tight junction loss.
Journal of clinical gastroenterology (2009-06-16)
Geertje Thuijls, Joep P M Derikx, Jacco-Juri de Haan, Joep Grootjans, Adriaan de Bruïne, Ad A M Masclee, Erik Heineman, Wim A Buurman
ABSTRACT
Tight junction breakdown, with loss of the important sealing protein claudin-3, is an early event in the development of intestinal damage. Therefore, noninvasive analysis of intestinal tight junction status could be helpful in early detection of intestinal injury. To investigate the usefulness of urinary claudin-3 as marker for intestinal tight junction loss. A rat hemorrhagic shock model and a human setting of known intestinal damage, that is, patients with relapsed inflammatory bowel disease (IBD), were used to investigate intestinal tight junction status by immunohistochemical staining and urinary claudin-3 levels by western blot. In rats claudin-3 urine levels increased rapidly after histologically proven intestinal tight junction loss, with significantly elevated levels at 90 minutes after shock compared with sham-operated animals [mean+/-SEM: 611+/-101 intensity (INT), n=6 vs. 232+/-30 INT, n=6; P<0.05]. Moreover, in colonic biopsies of patients with IBD relapse claudin-3 staining was reduced compared with biopsies of patients with IBD without signs of disease. Concomitantly, significantly increased claudin-3 urine levels were found in these patients (502+/-67 INT, n=10) compared with patients with IBD in remission (219+/-17 INT, n=10, P<0.001) and healthy volunteers (225+/-38 INT, n=10, P<0.001). Here we show for the first time in both an experimental and clinical setting a strong relation between intestinal tight junction loss and urinary claudin-3 levels. These findings suggest that measurement of urinary claudin-3 can be used as noninvasive marker for intestinal tight junction loss. This offers new opportunities for early diagnosis and follow-up of intestinal injury.