콘텐츠로 건너뛰기
Merck

다음으로 건너뛰기

941816

Magnesium chloride, hydrate

≥99.99% trace metals basis

greener alternative

더 친환경적인 대안

동의어(들):

Magnesium dichloride hydrate, Magnesium dichloride monohydrate

조직 및 계약 가격을 보려면 로그인를 클릭합니다.

크기 선택

보기 변경
사이즈/SKU재고 정보가격 (VAT 별도)
100 g
구입 가능 여부는 고객센터에 문의하십시오.
₩355,859
250 g
구입 가능 여부는 고객센터에 문의하십시오.
₩692,027

제품정보 (DICE 배송 시 비용 별도)

Linear Formula:
MgCl2
CAS 번호:
Molecular Weight:
95.21 (anhydrous basis)
NACRES:
NA.21
Assay:
(EDTA titration), ≥99.99% trace metals basis
Form:
powder or crystals
Solubility:
water: soluble

₩355,859


구입 가능 여부는 고객센터에 문의하십시오.

기술 서비스
도움이 필요하신가요? 저희 숙련된 과학자 팀이 도와드리겠습니다.
도움 문의


Quality Segment

assay

(EDTA titration), ≥99.99% trace metals basis

form

powder or crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100 ppm (metals basis)

mp

714 °C

solubility

water: soluble

density

2.32 g/cm3

anion traces

sulfate (SO42-): ≤50 ppm

cation traces

Al: ≤10 ppm, B: ≤10 ppm, Ba: ≤10 ppm, Ca: ≤10 ppm, Co: ≤10 ppm, Cr: ≤10 ppm, Cu: ≤10 ppm, Fe: ≤10 ppm, K: ≤10 ppm, Mn: ≤10 ppm, Na: ≤10 ppm, Ni: ≤10 ppm, Si: ≤10 ppm, Ti: ≤10 ppm, Zn: ≤10 ppm

greener alternative category

SMILES string

Cl[Mg]Cl.O

InChI

InChI=1S/2ClH.Mg.H2O/h2*1H;;1H2/q;;+2;/p-2

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product belongs to an Enabling category, and is a key component in greener battery technologies due to its abundance and lower environmental impact. It enhances battery safety and stability by reducing the risk of flammability and dendrite formation. Additionally, innovative electrolytes using magnesium chloride improve the efficiency and longevity of magnesium-ion batteries. Click here for more information.
Magnesium Chloride hydrate, is a white crystalline substance with properties of a Lewis acid. It dissolves in water. It is also slightly soluble in acetone and pyridine. Anhydrous MgCl2 is commonly used as a precursor in the production of metallic magnesium. OurMagnesium Chloride hydrate is suitable molten salt to prepare different types of eutectic mixtures due to its high operation temperature for thermal energy storage (TES) materials and heat transfer fluids (HTFs) for next-generation concentrating solar power (CSP) systems.

Application

Our >99.99% Magnesium Chloride hydrate, with low impurities, and solubility in water, is suitable for various applications. Example:Magnesium Chloride, is well-suited for use in magnesium batteries, particularly in the production of MgTFSI2/MgCl2 (DME) electrolyte solutions. These solutions have been found to effectively dissolve a substantial amount of magnesium chloride, resulting in enhanced electrochemical performance, notably in terms of reversibility.[] Other studies have indicated that the synthesis of MgTFSI2/MgCl2/DME electrolyte has led to the development of the first rechargeable Mg/S batteries with exceptional cycling stability, achieved by mitigating polysulfide dissolution. The battery is reported to primarily operate through S0/MgSx redox processes. [] It has been reported that the significance of Magnesium chloride, in the formulation of a concentrated Mg(HMDS)2/MgCl2 solution in THF electrolyte lies in its pivotal role in enabling a reverse Schlenk equilibrium. This unique preparation method facilitates the reversible deposition and dissolution of magnesium, broadening the electrochemical range and achieving an impressive coulombic efficiency of 99%. Moreover, the exceptional solubility of MgCl2 in THF, bolstered by the presence of Mg(HMDS)2, contributes to the electrolyte′s outstanding performance in terms of high reversible capacities and robust rate capabilities. This underscores the vital importance of MgCl2 in offering a novel approach for the advancement of magnesium electrolytes. []Furthermore,Magnesium chloride, is not only essential for batteries but also serves as a catalyst in olefin polymerization. It plays a critical role as a fundamental component in Ziegler-Natta catalysts employed for olefin polymerization. The presence of magnesium chloride is instrumental in facilitating the polymerization process, thereby enabling the production of diverse olefin-based polymers.[]

Our Magnesium Chloride hydrate, with a purity of ≥99.99%, is an excellent choice as a source for Magnesium electrolytes for batteries. Its properties and high purity make it highly suitable for Solar Energy Storage in Phase Change Materials for next-generation concentrating solar power (CSP) systems.[]

Other Notes

No metal contact and handle under the glove box


저장 등급

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



가장 최신 버전 중 하나를 선택하세요:

시험 성적서(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.

도움이 필요하시면 연락하세요. 고객 지원 부서

이 제품을 이미 가지고 계십니까?

문서 라이브러리에서 최근에 구매한 제품에 대한 문서를 찾아보세요.

문서 라이브러리 방문