form
powder
Quality Segment
feed ratio
lactide:glycolide 50:50
mol wt
PLGA average Mn 40,000, (PGlu average Mn 2,000)
color
white to beige
shipped in
dry ice
storage temp.
−20°C
General description
PLGA-b-PGA block copolymer is a innovative material engineered to accelarate drug delivery and biomedical research. This amphiphilic copolymer combines the hydrophobic, biodegradable properties of PLGA with the hydrophilic, biocompatible nature of poly(glutamic acid) (PGA), offering unmatched versatility for tailored therapeutic applications.
Application
This polymer is a amphiphilic diblock copolymer composed of a hydrophobic PLGA block and a hydrophilic poly(glutamic acid) block. PLGA is a biocompatible and biodegradable polymer that has been approved by the FDA and the EMA for biomedical and pharmaceutical applications. Poly(glutamic acid) is biodegradable, non-toxic and non-immunogenic macromolecules. This biodegradable, biocompatible polymer can self-assemble to form nanoparticles, such as micelles and polymersomes, for controlled release of molecules applications
1 of 1
이 품목 | |||
|---|---|---|---|
| description PLGA average Mn 40,000, mol wt (PGlu average Mn 2,000), lactide:glycolide 50:50 | description PLGA average Mn 25,000, mol wt (PGlu average Mn 5,000), lactide:glycolide 50:50 | description PEG average Mn 5,000, PLGA Mn 15,000 | description 10k-2k |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| color white to beige | color white to beige, powder | color - | color - |
| feed ratio lactide:glycolide 50:50 | feed ratio lactide:glycolide 50:50 | feed ratio - | feed ratio - |
| shipped in dry ice | shipped in - | shipped in - | shipped in - |
| mol wt PLGA average Mn 40,000, (PGlu average Mn 2,000) | mol wt PLGA average Mn 25,000, (PGlu average Mn 5,000) | mol wt PEG average Mn 5,000, PLGA Mn 15,000 | mol wt - |
| storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under Poly(D,L-lactic-co-glycolic acid)-block-poly(glutamic acid)
저장 등급
11 - Combustible Solids
인화점 (F)
Not applicable
인화점 (°C)
Not applicable



