packaging
pack of 1000 ea
manufacturer/tradename
Hirschmann 9211375
L × I.D. × O.D.
75 mm × 0.95 mm × 1.35 mm
tube diam.
0.95 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
1 of 4
이 품목 | Z611220 | Z611204 | Z611190 |
|---|---|---|---|
| manufacturer/tradename Hirschmann 9211375 | manufacturer/tradename Hirschmann 9211380 | manufacturer/tradename Hirschmann 9211310 | manufacturer/tradename Hirschmann 9211080 |
| packaging pack of 1000 ea | packaging pack of 1000 ea | packaging pack of 1000 ea | packaging pack of 1000 ea |
| L × I.D. × O.D. 75 mm × 0.95 mm × 1.35 mm | L × I.D. × O.D. 80 mm × 0.95 mm × 1.35 mm | L × I.D. × O.D. 100 mm × 0.95 mm × 1.35 mm | L × I.D. × O.D. 80 mm × 0.8 mm × 1 mm |
| tube diam. 0.95 mm | tube diam. 0.95 mm | tube diam. 0.95 mm | tube diam. 0.8 mm |
Legal Information
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.