material
polypropylene
무균
sterile
특징
DNA free (Human)
PCR clean
RNase and DNase free
autoclavable
endotoxin free
pyrogen free
제조업체/상표
991-00105
포장
pkg of 20 packs of 96 each
용적
200 μL
일반 설명
200 µL filtered pipette tips for the Opentrons Flex, provided sterile in racks or blister packs (refills). Tips are clear, non-conducting polypropylene tips, free of human DNA, PCR inhibitors, RNase/DNase, pyrogens, and endotoxins. Opentrons Flex Tips and Racks are autoclavable.
법적 정보
AAW is a trademark of Millipore Corporation
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Opentrons is a registered trademark of Opentrons Labworks, Inc.
시험 성적서(COA)
관련 콘텐츠
View an automated protocol for Duolink® assays on the AAW™ automated assay workstation and see results comparing manual vs automated runs.
Read an automated protocol for protein purification using PureProteome™ nickel magnetic beads on the AAW™ automated assay workstation and see results comparing manual vs automated runs.