Merck
99100300
Opentrons Flex Tough Auto-Sealing Lids
new
Opentrons Flex Tough Auto-Sealing Lids
Slide 1 of 1
로그인조직 및 계약 가격 보기
크기 선택
1 EACH
₩432,289
₩432,289
구입 가능 여부는 고객센터에 문의하십시오.
다음으로 건너뛰기
기술 서비스
도움이 필요하신가요? 저희 숙련된 과학자 팀이 도와드리겠습니다.
도움 문의
material
polysulfone
제조업체/상표
991-00300
포장
pkg of 20 count
호환성
Opentrons Flex Riser
일반 설명
OPENTRONSFLEX Tough Auto-Sealing Lids provide automatic sealing of PCR plates on deck and are perfect for workflows that either want to minimize cross contamination or evaporation during their thermocycler incubations. The lids can be placed directly on the deck or with the OPENTRONSFLEX Riser, which will raise the position of the lids on the deck and make them stackable.The lids are added and removed from PCR plates using the AAW™ Flex Gripper and the OPENTRONSFLEX Thermocycler to create the seal. Once the incubation is complete, the customer can remove and dispose of the thermocycler lid and add a new one if there is a subsequent incubation.
법적 정보
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.