특징
Smart sensors
포장
pack of 1 ea
제조업체/상표
999-00196
용량
5—1000 μL
일반 설명
The OPENTRONSFLEX™ 96-Channel pipette enables accurate high throughput pipetting, optimized for working in 96-well plates. Flex Pipettes use air displacement technology to offer highly accurate pipetting with pipette volume ranges from 5 – 1000 μL.Smart sensors support automatic calibration, real-time positioning, and error detection. The 96-channel pipette occupies both pipette mounts and can easily be removed and swapped to an alternate pipette configurations to adapt your workflow needs.Four 96-Channel Tip Rack Adapters are included with the purchase of one Flex 96-Channel Pipette.
법적 정보
AAW is a trademark of Millipore Corporation
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.