material
Barex® plate (co-polymer), Durapore PVDF membrane, white plate
sterility
sterile
usage
Automated
feature
hydrophilic
packaging
pkg of 10 plates
technique(s)
activity assay: suitable, bead-based assay: suitable, ligand binding assay: suitable, low protein binding filtration: suitable
plate size
96 wells
working volume
50-250 μL
pore size
0.45 μm pore size
application(s)
sample preparation
compatibility
centrifuge & manifold
Legal Information
Barex is a registered trademark of INEOS Barex AG