Product Name
Cadaverine purum, ≥97.0% (GC)
Product Number
33211
Product Brand
sigma
CAS Number
462-94-2
Molecular Weight
102.18
Test
Specification
APPEARANCE (COLOR)Colorless to Very Faint Yellow
APPEARANCE (FORM)Liquid
COLOR (IN APHA)≤ 50 APHA
TITRATION (T) HCL 1M97.0 - 103.0 %
CONTENT OF CARBONATE (T)≤ 0.2 %
PURITY (GC AREA %)≥ 97.0 %
REFRACTIVE INDEX N20/D1.457 - 1.459
WATER≤ 1.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD48 MONTHS