Skip to Content
Products
Cart
0
KZ
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Back
Isotopic certified reference material of vanillin
Synonym(s):
Isotopic certified reference material of vanillin
Products
Product Category
certified reference materials (CRM)
Manufacturer
Application
Special Grade
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
VANA-1
Isotopic certified reference material of vanillin,
certified reference material
Expand
View Pricing
VANB-1
Isotopic certified reference material of vanillin,
certified reference material
Expand
View Pricing