Merck
  • Polymer-functionalized single-walled carbon nanotubes as a novel sol-gel solid-phase micro-extraction coated fiber for determination of poly-brominated diphenyl ethers in water samples with gas chromatography-electron capture detection.
Polymer-functionalized single-walled carbon nanotubes as a novel sol-gel solid-phase micro-extraction coated fiber for determination of poly-brominated diphenyl ethers in water samples with gas chromatography-electron capture detection.
Analytical chemistry (2009-04-15)
Weiya Zhang, Yin Sun, Caiying Wu, Jun Xing, Jianying Li
ABSTRACT
Single-walled carbon nanotubes (SWNTs) were functionalized with a hydroxyl-terminated silicone oil (TSO-OH). It is synthesized by the reactions of carbonyl chloride groups on the surface of SWNTs and hydroxyl groups of silicone oil (TSO-OH). The functionalized product SWNTs-TSO-OH was first used as precursor and selective stationary phase to prepare the sol-gel derived poly(SWNTs-TSO-OH) solid-phase microextraction (SPME) fiber for determination of polybrominated diphenyl ethers (PBDEs) in water samples. The possible major reaction of the sol-gel coating process was discussed and confirmed by IR spectra, Raman spectroscopy, and scanning electron microscopy. Some parameters of SPME fiber for the determination of PBDEs were investigated by headspace SPME/gas chromatography with electron-capture detection (HS-SPME/GC-ECD). Compared with the commercial SPME fiber, the new coated fiber showed higher extraction efficiency to PBDEs, better thermal stability (over 340 degrees C), and longer life span (over 200 times). All of these advantages are mainly due to the incorporation of SWNTs, which enhanced the pi-pi interaction with PBDEs and increased the surface area of extraction in contact with the sample. Moreover, the sol-gel coating technology additionally provided the porous structure of the 3-D silica network and the strong chemical binding provided which also will improve the extraction efficiency. Under optimized conditions, the method detection limits for seven PBDEs were 0.08-0.8 ng/L (S/N = 3) and the precision (RSD, n = 5) was 2.2-7.5% at the 50 ng/L level. The linearity of the developed method is in the range of 5-500 ng/L with coefficients of correlation greater than 0.995. The developed method was successfully applied for the analysis of trace PBDEs in reservoir water and wastewater samples. The recoveries obtained at spiking 50 ng/L were between 74% and 109% (n = 5) for PBDEs in water samples.
Product Description
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1, plain gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, light blue hub
Supelco
SPME Overcoated Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, notched pink hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub