Merck
  • Biotin tagging of MeCP2 in mice reveals contextual insights into the Rett syndrome transcriptome.
Biotin tagging of MeCP2 in mice reveals contextual insights into the Rett syndrome transcriptome.
Nature medicine (2017-09-19)
Brian S Johnson, Ying-Tao Zhao, Maria Fasolino, Janine M Lamonica, Yoon Jung Kim, George Georgakilas, Kathleen H Wood, Daniel Bu, Yue Cui, Darren Goffin, Golnaz Vahedi, Tae Hoon Kim, Zhaolan Zhou
ABSTRACT
Mutations in MECP2 cause Rett syndrome (RTT), an X-linked neurological disorder characterized by regressive loss of neurodevelopmental milestones and acquired psychomotor deficits. However, the cellular heterogeneity of the brain impedes an understanding of how MECP2 mutations contribute to RTT. Here we developed a Cre-inducible method for cell-type-specific biotin tagging of MeCP2 in mice. Combining this approach with an allelic series of knock-in mice carrying frequent RTT-associated mutations (encoding T158M and R106W) enabled the selective profiling of RTT-associated nuclear transcriptomes in excitatory and inhibitory cortical neurons. We found that most gene-expression changes were largely specific to each RTT-associated mutation and cell type. Lowly expressed cell-type-enriched genes were preferentially disrupted by MeCP2 mutations, with upregulated and downregulated genes reflecting distinct functional categories. Subcellular RNA analysis in MeCP2-mutant neurons further revealed reductions in the nascent transcription of long genes and uncovered widespread post-transcriptional compensation at the cellular level. Finally, we overcame X-linked cellular mosaicism in female RTT models and identified distinct gene-expression changes between neighboring wild-type and mutant neurons, providing contextual insights into RTT etiology that support personalized therapeutic interventions.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) antibody produced in goat, whole antiserum
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Normal Rabbit IgG, This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.
Sigma-Aldrich
Anti-Parvalbumin Antibody, ascites fluid, clone PARV-19, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-MAP Kinase Kinase 4 (MKK4, SEK, JNKK1) (pThr223) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, Alexa Fluor488 conjugated, clone A60, Chemicon®, from mouse