Merck
  • Prostaglandin E2 suppresses hCAP18/LL-37 expression in human macrophages via EP2/EP4: implications for treatment of Mycobacterium tuberculosis infection.
Prostaglandin E2 suppresses hCAP18/LL-37 expression in human macrophages via EP2/EP4: implications for treatment of Mycobacterium tuberculosis infection.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-02-07)
Min Wan, Xiao Tang, Rokeya Sultana Rekha, S S V Jagadeeswara Rao Muvva, Susanna Brighenti, Birgitta Agerberth, Jesper Z Haeggström
ABSTRACT
Prostaglandin (PG)E2 is an arachidonic acid-derived lipid mediator that plays an important role in inflammation and immunity. In this study, we demonstrate that PGE2 suppresses basal and 1,25-dihydroxy vitamin D3 (VD3)-induced expression of hCAP18/LL-37 via E prostanoid (EP)2 and EP4 receptors. In humans, VD3 up-regulates vitamin D receptor (VDR) expression and promotes transcription of the cathelicidin hCAP18/LL-37 gene, whereas PGE2 counteracts this effect. We find that PGE2 induces the cAMP/PKA-signaling pathway and enhances the expression of the inhibitory transcription factor cAMP-responsive modulator/inducible cAMP early repressor, which prevents VDR expression and induction of hCAP18/LL-37 in human macrophages. The negative regulation by PGE2 was evident in M1- and M2-polarized human macrophages, although PGE2 displayed more profound inhibitory effects in M2 cells. PGE2 impaired VD3-induced expression of cathelicidin and concomitant activation of autophagy during Mycobacterium tuberculosis (Mtb) infection and facilitated intracellular Mtb growth in human macrophages. An EP4 agonist also significantly promoted Mtb survival in human macrophages. Our results indicate that PGE2 inhibits hCAP18/LL-37 expression, especially VD3-induced cathelicidin and autophagy, which may reduce host defense against Mtb. Accordingly, antagonists of EP4 may constitute a novel adjunctive therapy in Mtb infection.-Wan, M., Tang, X., Rekha, R. S., Muvva, S. S. V. J. R., Brighenti, S., Agerberth, B., Haeggström, J. Z. Prostaglandin E2 suppresses hCAP18/LL-37 expression in human macrophages via EP2/EP4: implications for treatment of Mycobacterium tuberculosis infection.
Sigma-Aldrich
TWEEN® 80, suitable for cell culture, suitable for insect cell culture, viscous liquid
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-100, laboratory grade