Merck
  • Development of QuEChERS Method for the Determination of Polycyclic Aromatic Hydrocarbons in Smoked Meat Products Using GC-MS from Qatar.
Development of QuEChERS Method for the Determination of Polycyclic Aromatic Hydrocarbons in Smoked Meat Products Using GC-MS from Qatar.
Journal of analytical methods in chemistry (2018-08-18)
Hussain Al-Thaiban, Nada Al-Tamimi, Murad Helaleh
ABSTRACT
A simple and fast method for the determination of PAHs in smoked meat samples was described. The QuEChERS (Z-Sep) procedure was used for sample preparation. Gas chromatograph-mass spectrometer with electron ionization (EI) was used to separate and detect the PAHs. All 16 common PAHs were analyzed successfully. Matrix-matched calibration was applied. Spiked samples were performed at 1 ng/g (n=10) and 10 ng/g (n=10) for two days. Overall recoveries of PAHs were within 74 to 117%, with RSDs within 1.15 to 37.57% and 1 and 10 ng/g wet weight for first and second day, respectively. In most of the analyzed smoked meat samples, there were no exceeded levels compared to the maximum levels declared by Commission Regulation (EU) number 835/2011. The method can be recommended for routine analysis for laboratories having a large number of samples.
Sigma-Aldrich
N-Acetyl-D-penicillamine, ≥99.0% (T)
Sigma-Aldrich
L-Pyroglutamic acid 2-naphthylamide, ≥99% (TLC)
Supelco
Supel QuE, Z-Sep+ Bulk, pkg of 20 g
Supelco
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100
Sigma-Aldrich
Acenaphthylene-d8, ≥98 atom % D, 98% (CP)
Supelco
Supel QuE, Z-Sep Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep/C18 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Bulk