Merck
  • BCL-XL binds and antagonizes RASSF6 tumor suppressor to suppress p53 expression.
BCL-XL binds and antagonizes RASSF6 tumor suppressor to suppress p53 expression.
Genes to cells : devoted to molecular & cellular mechanisms (2017-12-02)
Xiaoyin Xu, Hiroaki Iwasa, Shakhawoat Hossain, Aradhan Sarkar, Junichi Maruyama, Kyoko Arimoto-Matsuzaki, Yutaka Hata
ABSTRACT
RASSF6, a member of the tumor suppressor Ras-association domain family proteins, induces apoptosis in the caspase-dependent and caspase-independent manners. RASSF6 interacts with MDM2 and stabilizes p53. BCL-XL is a prosurvival member of BCL-2 family proteins. BCL-XL directly inhibits proapoptotic BAX and BAK. BCL-XL also traps tBID, a proapoptotic activator BH3-only protein, and sequesters p53. In addition, BCL-XL regulates the mitochondrial membrane permeability via voltage-dependent anion channel. In these manners, BCL-XL plays an antiapoptotic role. We report the interaction of BCL-XL with RASSF6. BCL-XL inhibits the interaction between RASSF6 and MDM2 and suppresses p53 expression. Consequently, BCL-XL antagonizes RASSF6-mediated apoptosis. Thus, the inhibition of RASSF6-mediated apoptosis also underlies the prosurvival role of BCL-XL.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Pepstatin A, microbial, Purity ≥98%