Merck
  • Phytosterol ester processing in the small intestine; impact on cholesterol availability for absorption and chylomicron cholesterol incorporation in healthy humans
Phytosterol ester processing in the small intestine; impact on cholesterol availability for absorption and chylomicron cholesterol incorporation in healthy humans
Journal of Lipid Research (2011)
Amiot MJ, et al.