Merck
  • Real-time three-dimensional tracking of single synaptic vesicles reveals that synaptic vesicles undergoing kiss-and-run fusion remain close to their original fusion site before reuse.
Real-time three-dimensional tracking of single synaptic vesicles reveals that synaptic vesicles undergoing kiss-and-run fusion remain close to their original fusion site before reuse.
Biochemical and biophysical research communications (2019-05-17)
Xianan Qin, Richard W Tsien, Hyokeun Park
ABSTRACT
The release of neurotransmitters via the fusion between synaptic vesicles and the presynaptic membrane is an essential step in synaptic transmission. Synaptic vesicles generally undergo two distinct modes of exocytosis called full-collapse fusion and kiss-and-run fusion. In kiss-and-run fusion, the fusion pore of the synaptic vesicle opens transiently without the vesicle collapsing fully into the plasma membrane; thus, each synaptic vesicle can be used multiple times to release neurotransmitters. Despite considerable research, the detailed mechanisms that underlie kiss-and-run fusion remain elusive, particularly the location of synaptic vesicles after kiss-and-run events. To address this question, we performed real-time three-dimensional tracking of single synaptic vesicles labeled with a single quantum dot in the presynaptic terminal of cultured hippocampal neurons and analyzed the three-dimensional trajectories of these vesicles undergoing kiss-and-run fusion. We found that the majority of these synaptic vesicles underwent another exocytosis event within 120 nm of their original fusion site and underwent a second exocytosis event within 10 s of the first fusion event. These results indicate that after kiss-and-run fusion, synaptic vesicles remain relatively close to their original fusion site and can release repeatedly at brief intervals, allowing neurons to maintain neurotransmitter release during bursting activity.
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Casein from bovine milk, powder