Merck
  • Engineered SUMO/protease system identifies Pdr6 as a bidirectional nuclear transport receptor.
Engineered SUMO/protease system identifies Pdr6 as a bidirectional nuclear transport receptor.
The Journal of cell biology (2019-04-27)
Arturo Vera Rodriguez, Steffen Frey, Dirk Görlich
ABSTRACT
Cleavage of affinity tags by specific proteases can be exploited for highly selective affinity chromatography. The SUMO/SENP1 system is the most efficient for such application but fails in eukaryotic expression because it cross-reacts with endogenous proteases. Using a novel selection system, we have evolved the SUMOEu/SENP1Eu pair to orthogonality with the yeast and animal enzymes. SUMOEu fusions therefore remain stable in eukaryotic cells. Likewise, overexpressing a SENP1Eu protease is nontoxic in yeast. We have used the SUMOEu system in an affinity-capture-proteolytic-release approach to identify interactors of the yeast importin Pdr6/Kap122. This revealed not only further nuclear import substrates such as Ubc9, but also Pil1, Lsp1, eIF5A, and eEF2 as RanGTP-dependent binders and thus as export cargoes. We confirmed that Pdr6 functions as an exportin in vivo and depletes eIF5A and eEF2 from cell nuclei. Thus, Pdr6 is a bidirectional nuclear transport receptor (i.e., a biportin) that shuttles distinct sets of cargoes in opposite directions.
Sigma-Aldrich
Monoclonal Anti-Maltose Binding Protein antibody produced in mouse, clone MBP-17, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins