Merck
  • The NH2-terminal region of focal adhesion kinase reconstitutes high affinity IgE receptor-induced secretion in mast cells.
The NH2-terminal region of focal adhesion kinase reconstitutes high affinity IgE receptor-induced secretion in mast cells.
The Journal of biological chemistry (2000-06-23)
D Vial, H Okazaki, R P Siraganian
ABSTRACT
Focal adhesion kinase (FAK) is tyrosine-phosphorylated by adherence of cells and also by FcepsilonRI aggregation in RBL-2H3 mast cells. Using phosphorylation site-specific antibodies, we observed that FcepsilonRI activation in these cells led to an increase in FAK phosphorylation at the same tyrosine residues that are phosphorylated by integrin-induced activation. Previous studies in the 3B6 line, a FAK-deficient variant of the RBL-2H3 cells, suggest that FAK plays a role in FcepsilonRI-induced secretion. Stable cell lines expressing either full-length or truncated forms of FAK were isolated after transfection of the FAK-deficient 3B6 variant cells. The NH(2) domain of FAK, which lacks the enzymatic and the COOH-terminal regions, was sufficient to reconstitute secretion. The different truncated forms of FAK were still tyrosine-phosphorylated after FcepsilonRI aggregation. Therefore, the kinase domain and the COOH-terminal region are not essential for FcepsilonRI-induced tyrosine phosphorylation of FAK or for secretion. Taken together, our data demonstrate that the reconstitution of secretion is dissociated from FAK activation and that the NH(2)-terminal region of FAK is the only critical element that may play a role in FcepsilonRI-induced secretion by acting as an adaptor or linker molecule.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse