Merck
  • Extracellular beta-nicotinamide adenine dinucleotide (beta-NAD) promotes the endothelial cell barrier integrity via PKA-and EPAC1/Rac1-dependent actin cytoskeleton rearrangement
Extracellular beta-nicotinamide adenine dinucleotide (beta-NAD) promotes the endothelial cell barrier integrity via PKA-and EPAC1/Rac1-dependent actin cytoskeleton rearrangement
Journal of Cellular Physiology (2010)
Umapathy NS, et al.