Merck
  • Electrochemical endotoxin aptasensor based on a metal-organic framework labeled analytical platform.
Electrochemical endotoxin aptasensor based on a metal-organic framework labeled analytical platform.
Materials science & engineering. C, Materials for biological applications (2020-01-12)
Yangyang Duan, Nan Wang, Zixu Huang, Hongxiu Dai, Ling Xu, Shengjun Sun, Houyi Ma, Meng Lin
ABSTRACT
An electrochemical aptasensor for the lipopolysaccharide (LPS) detection was constructed by using copper-based metal-organic framework (Cu-MOF) as a label and the LPS aptamer of specific single-stranded DNA as a probe. The carboxyl-functionalized polypyrrole nanowires (PPy NWs) were synthesized by electrochemical polymerization method, and the amino-terminated aptamer covalently coupling with the carboxyl group of the PPy NWs was immobilized onto the modified electrode. The aptamer can specifically combine with the target LPS molecules, and Cu-MOF was labeled by adsorption based on specific interactions of LPS carbohydrate portions with anionic groups. The fabrication processes of the aptasensor were characterized by scanning electron microscopy (SEM), X-ray photoelectron spectroscopy (XPS) and electrochemical impedance spectroscopy (EIS). Differential pulse voltammetry (DPV) was used to measure electrochemical performance of the aptasensor, and the electrochemical signal can be directly measured by the electrochemical redox reaction of Cu(II)/Cu(I) existed in the Cu-MOF. The electrochemical aptasensor exhibited a high sensitivity toward LPS ranging from 1.0 pg/mL to 1.0 ng/mL with a detection limit of 0.29 pg/mL. Moreover, the developed sensor was found to have good selectivity, stability and regeneration properties, and the sensor also successfully detected LPS in real tap water samples.
Sigma-Aldrich
Pyrrole-2-carboxylic acid, 99%