Merck
  • Cytotoxic effects of a chlorhexidine mouthwash and of an enzymatic mouthwash on human gingival fibroblasts.
Cytotoxic effects of a chlorhexidine mouthwash and of an enzymatic mouthwash on human gingival fibroblasts.
Odontology (2019-10-19)
Ana Sofia Coelho, Mafalda Laranjo, Ana Cristina Gonçalves, Anabela Paula, Siri Paulo, Ana Margarida Abrantes, Francisco Caramelo, Manuel Marques Ferreira, Mário Jorge Silva, Eunice Carrilho, Maria Filomena Botelho
ABSTRACT
The aim of this study was to evaluate the cytotoxic effects of an enzymatic mouthwash and of a chlorhexidine mouthwash on human gingival fibroblasts. The metabolic activity of the fibroblasts exposed to each mouthwash was assessed by the MTT assay and the protein content was assessed by the SRB assay. The flow cytometry was used to evaluate the cell cycle and the types of cell death. The oxidative status was evaluated through the DCF and the DHE probes and the intracellular GSH concentration and the mitochondrial membrane potential through JC-1. The cytotoxicity of both mouthwashes was found to be dependent on the exposure time and on the concentration. However, the cytotoxicity of the enzymatic mouthwash was found to be lower than that of the chlorhexidine mouthwash. A trend towards increased oxidative stress was observed for both mouthwashes. After exposing the fibroblasts to the mouthwashes, a G2/M phase block was observed and cell death occurred predominantly by necrosis. The effects of chlorhexidine on fibroblasts were identified at lower concentrations than those used in clinical practice. Therefore, the use of chlorhexidine as an antiseptic in surgical and postoperative situations should be limited. In order to clarify the clinical significance of the enzymatic mouthwash cytotoxicity new clinical studies will be necessary.
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium chloride, anhydrous, powder, ≥97%
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Methanol, anhydrous, 99.8%