Merck
  • Cas9-mediated allelic exchange repairs compound heterozygous recessive mutations in mice.
Cas9-mediated allelic exchange repairs compound heterozygous recessive mutations in mice.
Nature biotechnology (2018-08-14)
Dan Wang, Jia Li, Chun-Qing Song, Karen Tran, Haiwei Mou, Pei-Hsuan Wu, Phillip W L Tai, Craig A Mendonca, Lingzhi Ren, Blake Y Wang, Qin Su, Dominic J Gessler, Phillip D Zamore, Wen Xue, Guangping Gao
ABSTRACT
We report a genome-editing strategy to correct compound heterozygous mutations, a common genotype in patients with recessive genetic disorders. Adeno-associated viral vector delivery of Cas9 and guide RNA induces allelic exchange and rescues the disease phenotype in mouse models of hereditary tyrosinemia type I and mucopolysaccharidosis type I. This approach recombines non-mutated genetic information present in two heterozygous alleles into one functional allele without using donor DNA templates.
Sigma-Aldrich
D-Saccharic acid 1,4-lactone monohydrate, ≥98.0% (HPLC)
Sigma-Aldrich
KOD Hot Start Master Mix, Ready-to-use 2X mixture, containing KOD Hot Start DNA Polymerase, two monoclonal antibodies, ultrapure deoxynucleotides, and reaction buffer with MgSO4, optimized for convenient high fidelity PCR., ready-to-use solution, suitable for PCR