Merck
  • Zinc finger protein Wiz links G9a/GLP histone methyltransferases to the co-repressor molecule CtBP.
Zinc finger protein Wiz links G9a/GLP histone methyltransferases to the co-repressor molecule CtBP.
The Journal of biological chemistry (2006-05-17)
Jun Ueda, Makoto Tachibana, Tsuyoshi Ikura, Yoichi Shinkai
ABSTRACT
G9a is a SET-domain mammalian histone methyltransferase responsible for mono- and dimethylation of lysine 9 in histone H3 (H3K9) at euchromatic regions. Recently we reported that G9a forms a stoichiometric heteromeric complex with another SET-domain-containing molecule, GLP/Eu-HMTase1. Although G9a and GLP can independently methylate H3K9 in vitro, G9a/GLP heteromeric formation seems to be essential for their function as a euchromatic H3K9 methyltransferase in vivo. To further elucidate how G9a/GLP-mediated histone methylation and transcriptional regulation are controlled, we purified and characterized G9a complexes from mouse embryonic stem cells. We identified a novel G9a/GLP-associating zinc finger molecule named Wiz that can interact with G9a and GLP independently but is more stable in the G9a/GLP heteromeric complexes. Interestingly, Wiz small inhibitory RNA knocks down not only Wiz but also G9a. GLP deficiency also decreases G9a levels, suggesting that the Wiz/G9a/GLP tri-complex may protect G9a from degradation and that Wiz plays a major role in G9a/GLP heterodimer formation. Furthermore, amino acid sequence analysis of Wiz predicted two potential CtBP binding sites, and indeed CtBP binding to Wiz and association of CtBP with the Wiz/G9a/GLP complex was observed. These data indicate that Wiz not only contributes to the stability of G9a but also links the G9a/GLP heteromeric complex to the CtBP co-repressor machinery.
Sigma-Aldrich
EHMT1 human, recombinant, expressed in E. coli, ≥63% (SDS-PAGE)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®