Merck
  • Tumor Necrosis Factor-Related Apoptosis-Inducing Ligand Receptor Deficiency Promotes the Ductular Reaction, Macrophage Accumulation, and Hepatic Fibrosis in the Abcb4-/- Mouse.
Tumor Necrosis Factor-Related Apoptosis-Inducing Ligand Receptor Deficiency Promotes the Ductular Reaction, Macrophage Accumulation, and Hepatic Fibrosis in the Abcb4-/- Mouse.
The American journal of pathology (2020-04-03)
Anuradha Krishnan, Tomohiro Katsumi, Maria E Guicciardi, Adiba I Azad, Nazli B Ozturk, Christy E Trussoni, Gregory J Gores
ABSTRACT
The tumor necrosis factor-related apoptosis-inducing ligand (TRAIL; TNFSF10) receptor (TR) is a pro-apoptotic receptor whose contribution to chronic cholestatic liver disease is unclear. Herein, we examined TRAIL receptor signaling in a mouse model of cholestatic liver injury. TRAIL receptor-deficient (Tnsf10 or Tr-/-) mice were crossbred with ATP binding cassette subfamily B member 4-deficient (Abcb4-/-, alias Mdr2-/-) mice. Male and female wild-type, Tr-/-, Mdr2-/-, and Tr-/-Mdr2-/- mice were assessed for liver injury, fibrosis, and ductular reactive (DR) cells. Macrophage subsets were examined by high-dimensional mass cytometry (time-of-flight mass cytometry). Mdr2-/- and Tr-/-Mdr2-/- mice had elevated liver weights and serum alanine transferase values. However, fibrosis was primarily periductular in Mdr2-/- mice, compared with extensive bridging fibrosis in Tr-/-Mdr2-/- mice. DR cell population was greatly expanded in the Tr-/-Mdr2-/- versus Mdr2-/- mice. The expanded DR cell population in Tr-/-Mdr2-/- mice was due to decreased cell loss by apoptosis and not enhanced proliferation. As assessed by time-of-flight mass cytometry, total macrophages were more abundant in Tr-/-Mdr2-/- versus Mdr2-/- mice, suggesting the DR cell population promotes macrophage-associated hepatic inflammation. Inhibition of monocyte-derived recruited macrophages using the CCR2/CCR5 antagonist cenicriviroc in the Mdr2-/- mice resulted in further expansion of the DR cell population. In conclusion, genetic deletion of TRAIL receptor increased the DR cell population, macrophage accumulation, and hepatic fibrosis in the Mdr2-/- model of cholestasis.
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
Direct Red 80, Dye content 25 %
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)