Merck
  • Genome-wide profiling of methylated promoters in pancreatic adenocarcinoma.
Genome-wide profiling of methylated promoters in pancreatic adenocarcinoma.
Cancer biology & therapy (2008-06-07)
Noriyuki Omura, Chung-Pin Li, Ang Li, Seung-Mo Hong, Kimberly Walter, Antonio Jimeno, Manuel Hidalgo, Michael Goggins
ABSTRACT
Many genes undergo aberrant methylation in human cancers, and microarray platforms enable more comprehensive profiling of aberrant DNA methylation patterns. 1,010 of 87,922 probes on the 88 K promoter array (606 genes) had a higher signal (log(2) > 2) in the pancreatic cancer line, Panc-1 compared to the non-neoplastic pancreatic duct line, HPDE. Using this cut-off, bisulfite sequencing and/or MSP confirmed differential methylation of all 27 genes (66 probes) predicted to be methylated by the MCA array. More than 1/2 of the genes aberrantly hypermethylated in Panc-1 were not expressed in the pancreatic duct (HPDE) by expression array analysis. Using the 244 K CpG island array, 1,968 CpG islands were differentially methylated in MiaPaca2 compared to normal pancreas. The MCA method was more likely to identify hypermethylation within CpG islands than a cocktail of methylation sensitive restriction enzymes. DNA methylation profiles using 10 ng of DNA were highly correlated with those obtained using 5 ug of DNA (R2 = 0.98). Analysis of 57 pancreatic cancers and 34 normal pancreata using MSP identified MDFI, hsa-miR-9-1, ZNF415, CNTNAP2 and ELOVL4 as methylated in 96%, 89%, 86%, 82% and 68% of the cancers vs. 9%, 15%, 6%, 3% and 97% of normal pancreata, respectively. We used methylated CpG island amplification (MCA) and Agilent promoter and CpG island microarrays to identify differential DNA methylation patterns in pancreatic cancer vs. normal pancreas. We examined MCA array reproducibility, compared it to methylation profiles obtained using a cocktail of methylation-sensitive restriction enzymes and examined gene expression of methylated genes. Promoter and CpG island array analysis finds aberrant methylation of hundreds of promoters and CpG islands in pancreatic cancer cells.
Sigma-Aldrich
Sodium bisulfite, JIS special grade, 64.0-67.4% SO2 basis
Sigma-Aldrich
Sodium bisulfite, anhydrous, free-flowing, Redi-Dri, ACS reagent
Sigma-Aldrich
Sodium bisulfite, ACS reagent