Merck
Anti-babesial and anti-plasmodial compounds from Phyllanthus niruri.
Journal of natural products (2005-04-23)
Subeki Subeki, Hideyuki Matsuura, Kosaku Takahashi, Masahiro Yamasaki, Osamu Yamato, Yoshimitsu Maede, Ken Katakura, Sumiko Kobayashi, Trimurningsih Trimurningsih, Chairul Chairul, Teruhiko Yoshihara
ABSTRACT
Bioassay-guided fractionation of boiled aqueous extracts from the whole plant of Phyllanthus niruri led to the isolation of 1-O-galloyl-6-O-luteoyl-alpha-d-glucose (1), with IC(50) values of 4.7 microg/mL against Babesia gibsoni and 1.4 microg/mL against Plasmodium falciparum in vitro. The known compounds beta-glucogallin (2), quercetin 3-O-beta-d-glucopyranosyl-(2-->1)-O-beta-d-xylopyranoside (3), beta-sitosterol, and gallic acid were also isolated. Structures of these compounds were elucidated on the basis of their chemical and spectroscopic data.
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)