Merck
  • The adverse effects of selenomethionine on skeletal muscle, liver, and brain in the steelhead trout (Oncorhynchus mykiss).
The adverse effects of selenomethionine on skeletal muscle, liver, and brain in the steelhead trout (Oncorhynchus mykiss).
Environmental toxicology and pharmacology (2020-07-01)
Jang-Won Lee, Dong-Fang Deng, Jinsu Lee, Kiyoung Kim, Hyun Jin Jung, Youngshik Choe, Seung Hwa Park, Minjung Yoon
ABSTRACT
Juvenile Oncorhynchus mykiss (average weight: 22.3 g) were fed one of five selenomethionine diets (1.09, 8.79, 15.37, 30.79, or 61.58 mg Se/kg diet). After 4 weeks, hepatic catalase activity over 15.37 mg Se/kg diets was significantly decreased, and the glutathione peroxidase activity over 30.79 mg Se/kg diets was elevated compared to the controls. In the brain, the dopamine levels at 61.58 mg Se/kg diet and the serotonin levels over 15.37 mg Se/kg diets were significantly increased, whereas the 3,4-dihydroxyphenylacetic acid, homovanillic acid, and dopamine turnover, and the 5-hydroxyindoleacetic acid and serotonin turnover over 30.79 mg Se/kg diets were decreased. In muscle, the 3-nitrotyrosine level over 15.37 mg Se/kg diets, acetylcholine esterase activity over 30.79 mg Se/kg diets, and histological alterations over 8.79 mg Se/kg diets were increased. Our current results showed that selenomethionine disrupted dopamine and serotonin metabolism in the brain and damaged the neuromuscular system in skeletal muscle.
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Supelco
3,4-Dihydroxyphenylacetic acid, analytical standard
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
5-Hydroxyindole-3-acetic acid, ≥98% (HPLC), crystalline
Sigma-Aldrich
Serotonin hydrochloride, powder