Merck
  • A mechanistic model to study the effect of the cell wall on starch digestion in intact cotyledon cells.
A mechanistic model to study the effect of the cell wall on starch digestion in intact cotyledon cells.
Carbohydrate polymers (2020-12-07)
Ana M Rovalino-Córdova, Victor Aguirre Montesdeoca, Edoardo Capuano
ABSTRACT
The role of the plant matrix is recognized as the main factor restricting starch digestibility in beans. Several authors have provided insights about the mechanisms behind the reduced starch digestibility in plant matrices. In this study, by means of a mathematical model, we provide a mechanistic explanation of the role played by the cell wall. It was confirmed that starch entrapped within intact cells could only be hydrolysed after α-amylase diffusion through the cell wall. This process is limited by the pores naturally present in the cell wall and the adsorption of α-amylase to the cell wall surface. These factors restrict the concentration of α-amylase available within the cells. The model assumptions are valid under controlled laboratory conditions and were validated with in-vitro digestion data giving very accurate results. The proposed approach provides new information to understand the digestibility of starch, and possibly other macronutrients, in complex food matrices.
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)