Merck
  • Interleukin-1beta up-regulates expression of neurofilament light in human neuronal cells.
Interleukin-1beta up-regulates expression of neurofilament light in human neuronal cells.
Journal of neurochemistry (2001-08-03)
H Rempel, L Kusdra, L Pulliam
ABSTRACT
Elevated expression of interleukin-1 (IL-1beta), a pro-inflammatory cytokine secreted by activated microglia, is a pathogenic marker of numerous neurodegenerative processes including Alzheimer's disease (AD). We have characterized a link between IL-1beta and the 68-kDa neurofilament light (NF-L) protein, which is a major component of the neuronal cytoskeleton. Using human brain aggregate cultures, we found that IL-1beta treatment significantly increased NF-L expression in primary neurons. Analysis of mRNA levels demonstrated elevated NF-L expression within 72 h while imaging of neurons by immunofluorescent staining for NF-L confirmed IL-1beta-induced NF-L protein expression. These observations suggest a potential inflammatory-induced mechanism for deregulation of an important cytoskeletal protein, NF-L, possibly leading to neuronal dysfunction.
Sigma-Aldrich
Anti-Neurofilament 200 kDa Antibody, clone N52, clone N52, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neurofilament 70 kDa Antibody, clone DA2, culture supernatant, clone DA2, Chemicon®