Merck
  • (3R,5S,7as)-(3,5-Bis(4-fluorophenyl)tetrahydro-1H-oxazolo[3,4-c]oxazol-7a-yl)methanol, a novel neuroprotective agent.
(3R,5S,7as)-(3,5-Bis(4-fluorophenyl)tetrahydro-1H-oxazolo[3,4-c]oxazol-7a-yl)methanol, a novel neuroprotective agent.
Journal of medicinal chemistry (2009-09-05)
Kelly E Desino, Sabah Ansar, Gunda I Georg, Richard H Himes, Mary Lou Michaelis, Douglas R Powell, Emily A Reiff, Hanumaiah Telikepalli, Kenneth L Audus
ABSTRACT
Compounds that interact with microtubules, such as paclitaxel, have been shown to possess protective properties against beta-amyloid (Abeta) induced neurodegeneration associated with Alzheimer's disease. In this work, the novel agent (3R,5S,7as)-(3,5-bis(4-fluorophenyl)tetrahydro-1H-oxazolo[3,4-c]oxazol-7a-yl)methanol was investigated for effectiveness in protecting neurons against several toxic stimuli and its interaction with the microtubule network. Exposure of neuronal cultures to Abeta peptide in the presence of 5 nM (3R,5S,7as)-(3,5-bis(4-fluorophenyl)tetrahydro-1H-oxazolo[3,4-c]oxazol-7a-yl)methanol resulted in a 50% increase in survival. Neuronal cultures treated with other toxic stimuli such as staurosporine, thapsigargin, paraquat, and H(2)O(2) showed significantly enhanced survival in the presence of (3R,5S,7as)-(3,5-bis(4-fluorophenyl)tetrahydro-1H-oxazolo[3,4-c]oxazol-7a-yl)methanol. Microtubule binding and tubulin assembly studies revealed differences compared to paclitaxel but confirmed the interaction of (3R,5S,7as)-(3,5-bis(4-fluorophenyl)tetrahydro-1H-oxazolo[3,4-c]oxazol-7a-yl)methanol with microtubules. Furthermore, in vitro studies using bovine brain microvessel endothelial cells experiments suggest that (3R,5S,7as)-(3,5-bis(4-fluorophenyl)tetrahydro-1H-oxazolo[3,4-c]oxazol-7a-yl)methanol can readily cross the blood-brain barrier in a passive manner.
Product Description
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)