Merck
  • Benzophenone-N-ethyl piperidine ether analogues--synthesis and efficacy as anti-inflammatory agent.
Benzophenone-N-ethyl piperidine ether analogues--synthesis and efficacy as anti-inflammatory agent.
Bioorganic & medicinal chemistry letters (2009-03-11)
Shaukath A Khanum, V Girish, S S Suparshwa, Noor Fatima Khanum
ABSTRACT
A sequence of substituted benzophenone-N-ethyl piperidine ether analogues has been synthesized and evaluated as orally active anti-inflammatory agents with reduced side effects. The anti-inflammatory and ulcerogenic activities of the compounds were compared with naproxen, indomethacin, and phenylbutazone. These analogues showed an interesting anti-inflammatory activity in carrageenan-induced foot pad edema assay. In the air-pouch test, some of the analogues reduced the total number of leukocytes of the exudate, which indicates inhibition of prostaglandin production. Side effects of the compounds were examined on gastric mucosa, in the liver and stomach. None of the compounds illustrated significant side effects compared with standard drugs like indomethacin and naproxen.
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Phenylbutazone
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications