Merck
  • KPC2 relocalizes HOXA2 to the cytoplasm and decreases its transcriptional activity.
KPC2 relocalizes HOXA2 to the cytoplasm and decreases its transcriptional activity.
Biochimica et biophysica acta (2015-08-26)
Laure Bridoux, Isabelle Bergiers, Amandine Draime, Mathias Halbout, Noémie Deneyer, Jean-Claude Twizere, René Rezsohazy
ABSTRACT
Regulation of transcription factor activity relies on molecular interactions or enzymatic modifications which influence their interaction with DNA cis-regulatory sequences, their transcriptional activation or repression, and stability or intracellular distribution of these proteins. Regarding the well-conserved Hox protein family, a restricted number of activity regulators have been highlighted thus far. In the framework of a proteome-wide screening aiming at identifying proteins interacting with Hoxa2, KPC2, an adapter protein constitutive of the KPC ubiquitin-ligase complex, was identified. In this work, KPC2 was confirmed as being a genuine interactor of Hoxa2 by co-precipitation and bimolecular fluorescence complementation assays. At functional level, KPC2 diminishes the transcriptional activity and induces the nuclear exit of Hoxa2. Gene expression analyses revealed that Kpc2 is active in restricted areas of the developing mouse embryo which overlap with the Hoxa2 expression domain. Together, our data support that KPC2 regulates Hoxa2 by promoting its relocation to the cytoplasm.
Sigma-Aldrich
Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Glutathione-S-Transferase (GST) antibody produced in mouse, clone GST-2, ascites fluid
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).