Merck
  • Fetuin-A is a HIF target that safeguards tissue integrity during hypoxic stress.
Fetuin-A is a HIF target that safeguards tissue integrity during hypoxic stress.
Nature communications (2021-01-24)
Stefan Rudloff, Mathilde Janot, Stephane Rodriguez, Kevin Dessalle, Willi Jahnen-Dechent, Uyen Huynh-Do
ABSTRACT
Intrauterine growth restriction (IUGR) is associated with reduced kidney size at birth, accelerated renal function decline, and increased risk for chronic kidney and cardiovascular diseases in adults. Precise mechanisms underlying fetal programming of adult diseases remain largely elusive and warrant extensive investigation. Setting up a mouse model of hypoxia-induced IUGR, fetal adaptations at mRNA, protein and cellular levels, and their long-term functional consequences are characterized, using the kidney as a readout. Here, we identify fetuin-A as an evolutionary conserved HIF target gene, and further investigate its role using fetuin-A KO animals and an adult model of ischemia-reperfusion injury. Beyond its role as systemic calcification inhibitor, fetuin-A emerges as a multifaceted protective factor that locally counteracts calcification, modulates macrophage polarization, and attenuates inflammation and fibrosis, thus preserving kidney function. Our study paves the way to therapeutic approaches mitigating mineral stress-induced inflammation and damage, principally applicable to all soft tissues.
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Sigma-Aldrich
Fetuin from fetal bovine serum, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Human AHSG / Alpha-2-HS-glycoprotein ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
Harris Hematoxylin Solution, Modified
Sigma-Aldrich
Direct Red 80, Dye content 25 %
Sigma-Aldrich
Eukitt® Quick-hardening mounting medium
Sigma-Aldrich
Soda lime, p.a., with indicator, pellets
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep