Merck
Cytotoxic triterpenoids from the stems of Microtropis japonica.
Journal of natural products (2009-06-19)
I-Hsiao Chen, Mei-Chin Lu, Ying-Chi Du, Ming-Hong Yen, Chin-Chung Wu, Yung-Husan Chen, Chih-Sheng Hung, Shu-Li Chen, Fang-Rong Chang, Yang-Chang Wu
ABSTRACT
Bioassay-guided fractionation of a methanol extract obtained from stems of Microtropis japonica led to the isolation of six new ursane-type triterpenoids (1-6) and a new 2,3-seco-oleanane-type triterpenoid (7), together with seven known compounds. The structures of the new compounds were elucidated using spectroscopic data analysis. Among the known compounds isolated, the main component, 8 (ursolic acid), was active for HL60 cells, and its effects on histone hyperacetylation and the inhibition of histone deacetylase (HDAC) activity were investigated.
Sigma-Aldrich
Ursolic acid, ≥90%
Supelco
Ursolic acid, analytical standard