Merck
  • Synthesis and biological evaluation of 4-piperidinecarboxylate and 4-piperidinecyanide derivatives for T-type calcium channel blockers.
Synthesis and biological evaluation of 4-piperidinecarboxylate and 4-piperidinecyanide derivatives for T-type calcium channel blockers.
Bioorganic & medicinal chemistry letters (2011-08-17)
Hyun Min Woo, Yun Suk Lee, Eun Joo Roh, Seon Hee Seo, Chi Man Song, Hye Jin Chung, Ae Nim Pae, Kye Jung Shin
ABSTRACT
To obtain selective and potent inhibitor for T-type calcium channel by ligand based drug design, 4-piperidinecarboxylate and 4-piperidinecyanide derivatives were prepared and evaluated for in vitro and in vivo activity against α(1G) calcium channel. Among them, several compounds showed good T-type calcium channel inhibitory activity and minimal off-target activity over hERG channel (% inhibition at 10 μM=61.85-71.99, hERG channel IC(50)=1.57 ± 0.14-4.98 ± 0.36 μM). Selected compound 31a was evaluated on SNL model of neuropathic pain and showed inhibitory effect on mechanical allodynia.
Sigma-Aldrich
Mibefradil dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Gabapentin, solid