Merck
  • Syntheses and characterization of nimesulide derivatives for dual enzyme inhibitors of both cyclooxygenase-1/2 and 5-lipoxygenase.
Syntheses and characterization of nimesulide derivatives for dual enzyme inhibitors of both cyclooxygenase-1/2 and 5-lipoxygenase.
Bioorganic & medicinal chemistry (2011-02-26)
Yue Li, Shu-Han Chen, Tian-Miao Ou, Jia-Heng Tan, Ding Li, Lian-Quan Gu, Zhi-Shu Huang
ABSTRACT
Cyclooxygenase-1/2 (COX-1/2) and 5-lipoxygenase (5-LOX) are enzymes in two different pathways in the inflammatory process. In the present study, a variety of new nimesulide derivatives were synthesized through incorporation of a 5-LOX pharmacophore into nimesulide followed with some structural modifications, which were then characterized for dual enzyme inhibitors for these two types of enzymes. Their structure-activity relationships (SARs) were studied, and compound 20f was found to be an excellent dual enzyme inhibitor. Its binding conformation and interaction mode were studied with molecular docking experiments. Compound 20f could become a lead compound for further development for potential anti-inflammatory drugs.
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)