Merck
  • Novel imidazo[2,1-b][1,3,4]thiadiazole carrying rhodanine-3-acetic acid as potential antitubercular agents.
Novel imidazo[2,1-b][1,3,4]thiadiazole carrying rhodanine-3-acetic acid as potential antitubercular agents.
Bioorganic & medicinal chemistry letters (2012-02-14)
Shankar G Alegaon, Kallanagouda R Alagawadi, Pranali V Sonkusare, Sagar M Chaudhary, Dilip H Dadwe, Amol S Shah
ABSTRACT
The increase in the prevalence of multi drug-resistant and extensively drug-resistant strains of Mycobacteriumtuberculosis case demonstrates the urgent need of discovering new promising compounds with antimycobacterial activity. As part of our research program and with a aim of identifying new antitubercular drug candidates, a new class of 2-(trifluoromethyl)-6-arylimidazo[2,1-b][1,3,4]thiadiazole derivatives has been synthesized by both conventional as well as microwave assisted method and evaluated for their in vitro antitubercular activity against M. tuberculosis H(37)Rv. Moreover, various drug-likeness properties of new compounds were predicted. Seven compounds from the series exhibited good activity with MIC in range 3.12-1.56μg/ml. The present study suggests that compounds 6b, 6c, 6d, 6e and 6f may serve as promising lead scaffolds for further generation of new anti-TB agents.
Product Description
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals