Merck
  • Synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives as platelet aggregation inhibitors.
Synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives as platelet aggregation inhibitors.
Bioorganic & medicinal chemistry letters (2011-12-06)
Xiao Tian, Li-Ying Wang, Shuai Xia, Zhu-Bo Li, Xing-Hui Liu, Yuan Yuan, Liang Fang, Hua Zuo
ABSTRACT
Novel 2H-benzo[b][1,4]oxazin-3(4H)-ones have been synthesized by condensation, reduction, O-alkylation and Smiles rearrangement using 3-bromo-4-hydroxy benzaldehyde, anilines, and chloroacetyl chloride as starting materials. All the synthesized compounds have been characterized by (1)H NMR, (13)C NMR, and HRMS, and tested for the inhibitory ability on platelet aggregation. The results have shown that the ADP (adenosine 5'-diphosphate)-induced platelet aggregation was inhibited by 7a-g with the IC(50) value at 10.14-18.83 μmol/L. Compound 7a exhibited the most potent inhibitory effect (IC(50)=10.14 μmol/L) among all the compounds, but less potent than the control drug ticlopidine (3.18 μmol/L) and aspirin (6.07 μmol/L). The preliminary structure-activity relationship (SAR) was initially investigated in the study.
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Aspirin, meets USP testing specifications