Merck
  • SLX4-XPF mediates DNA damage responses to replication stress induced by DNA-protein interactions.
SLX4-XPF mediates DNA damage responses to replication stress induced by DNA-protein interactions.
The Journal of cell biology (2020-12-22)
Riko Ishimoto, Yota Tsuzuki, Tomoki Matsumura, Seiichiro Kurashige, Kouki Enokitani, Koki Narimatsu, Mitsunori Higa, Nozomi Sugimoto, Kazumasa Yoshida, Masatoshi Fujita
ABSTRACT
The DNA damage response (DDR) has a critical role in the maintenance of genomic integrity during chromosome replication. However, responses to replication stress evoked by tight DNA-protein complexes have not been fully elucidated. Here, we used bacterial LacI protein binding to lacO arrays to make site-specific replication fork barriers on the human chromosome. These barriers induced the accumulation of single-stranded DNA (ssDNA) and various DDR proteins at the lacO site. SLX4-XPF functioned as an upstream factor for the accumulation of DDR proteins, and consequently, ATR and FANCD2 were interdependently recruited. Moreover, LacI binding in S phase caused underreplication and abnormal mitotic segregation of the lacO arrays. Finally, we show that the SLX4-ATR axis represses the anaphase abnormality induced by LacI binding. Our results outline a long-term process by which human cells manage nucleoprotein obstacles ahead of the replication fork to prevent chromosomal instability.
Sigma-Aldrich
IgG, Goat, Goat IgG Polyclonal Antibody control validated for use in ELISA.
Sigma-Aldrich
Anti-SLX1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PICH Antibody, clone 142-26-3, clone 142-26-3, from mouse